missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Herasafe™ 2030i Biological Safety Cabinets
GREENER_CHOICE

Herasafe 2030i BASIC, unconfigured BSC to EN12469, 230V, 50Hz. Herasafe 2030i CONFIGURED to include Cross Beam Smart UV. Additional safety features for ultimate protection and graphic user interface for ease of operation.

128970.16 DKK - 185207.15 DKK

Specifications

Certifications/Compliance EN 12469
Chamber Material Stainless Steel (316)
Product Type Biosafety Cabinet
Model Herasafe 2030i
Sash Opening (Metric) 200 mm
View More Specs

Products 8
Product Code Brand Description Dimensions (L x W x H) Exterior Outlets Price Quantity & Availability  
Product Code Brand Description Dimensions (L x W x H) Exterior Outlets Price Quantity & Availability  
15943120
GREENER_CHOICE
View accessory products View Documents
Thermo Scientific™
51032324
Herasafe 2030i, 0.9 m, with 200 mm Sash Opening, No options. 800 x 1000 x 1536 2 Rear Wall
128970.16 DKK
Each
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15953120
GREENER_CHOICE
View accessory products View Documents Promotion Details
Thermo Scientific™
51032325
Herasafe 2030i, 1.2 m, with 200 mm Sash Opening, No options. 800 x 1300 x 1536 2 Rear Wall
134218.95 DKK
Each
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15963120
GREENER_CHOICE
View accessory products View Documents Promotion Details
Thermo Scientific™
51032326
Herasafe 2030i, 1.5 m, with 200 mm Sash Opening, No options. 800 x 1600 x 1536 4 Rear Wall
167961.15 DKK
Each
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15973120
GREENER_CHOICE
View accessory products View Documents Promotion Details
Thermo Scientific™
51032327
Herasafe 2030i, 1.8 m, with 200 mm Sash Opening, No options. 800 x 1900 x 1536 4 Rear Wall
172460.10 DKK
Each
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15983120
GREENER_CHOICE
View Documents
Thermo Scientific™
51032328
Herasafe 2030i, 0.9 m, with 200 mm Sash Opening, Cross Beam UV in Side Walls. 800 x 1000 x 1536 2 Rear Wall
141717.21 DKK
Each
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15993120
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
51032329
Herasafe 2030i, 1.2 m, with 200 mm Sash Opening, Cross Beam UV in Side Walls. 800 x 1300 x 1536 2 Rear Wall
146966.00 DKK
Each
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17800325
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
51032330
Herasafe 2030i, 1.5 m, with 200 mm Sash Opening, Cross Beam UV in Side Walls. 800 x 1600 x 1536 4 Rear Wall
180708.20 DKK
Each
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15913130
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
51032331
Herasafe 2030i, 1.8 m, with 200 mm Sash Opening, Cross Beam UV in Side Walls. 800 x 1900 x 1536 4 Rear Wall
185207.15 DKK
Each
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Thermo Scientific™ Herasafe™ 2030i Biological Safety Cabinets (BSC) are designed to maximize sample protection and user safety with exceptional containment measures, while at the same time, delivering excellent user comfort and convenience. Herasafe 2030i biosafety cabinets incorporate innovative features, such as SmartFlow Plus, DAVe Plus and Smart UV technologies for excellent protection. Alarms, alerts, system self-checks and performance status are provided on a full-color touchscreen. With enabled connectivity (available in select countries) the remote app provides all data you need.

Breakthrough innovations and unprecedented features make the Herasafe 2030i BSC our most technologically advanced and user-friendly Class II, Type A2 cabinet available.

Smart protection:

  • Thermo Scientific™ SmartFlow™ Plus Technology - Autonomous safety features maintain proper air flow and monitor critical conditions in real time without user interaction.
  • Self-adjusting germicidal UV light intensity - Our unique cross beam UV system has exclusive self-adjusting exposure capability to enable reproducible germicidal effectiveness with every use.
  • 316L stainless steel interior - robust and resistant to scratches and chemicals

Simplified operation and ergonomics:

  • Touchscreen interface
  • SmartClean Plus window design – Easily access the interior of the BSC with the motorized front sash which automatically stops at the set working height. The sash can be further opened electronically or the whole front panel hinges open for thorough cleaning access.
  • Bright work area with glass side walls and LED lights
  • Ergonomic Arm Rests (option)
  • SmartPort convenience - Easily pass tubing and wiring through sidewall to accommodate many applications needs.

Information management:

  • Touchscreen and connectivity (available in select countries) – Built-in WiFi system is ready for cloud-based capabilities like remote equipment monitoring, secure data storage, collaboration tools and more. Digital Airflow Verification (DAVe) alerts, system self-checks and performance status are provided on the full-color touchscreen.

Energy efficiency:

  • LED interior illumination
  • Dual DC motors
  • Standby mode

Certifications:

  • For North America, cabinets are NSF-49 certified (4ft and 6ft models)
  • For Europe, the cabinets are certified according to EN12469 (all 4 sizes). The 1.2 m model has been certified to EN 12469 with the NF mark by LNE as well.

Available sizes:

  • North America 4ft and 6ft models, other countries: 0.9 m, 1.2 m, 1.5 m and 1.8 m

For cell and gene therapy applications, we offer the Herasafe 2030i Biological Safety Cabinet CTS (Cell Therapy Systems) Series which consists of the product, a comprehensive documentation package including certifications and protocols, as well as compliance services.

Previous models: Herasafe KS

Note: Please order stand separately and indicate any additional requirements/ options you need when you Request a quote

Specifications

Specifications

EN 12469
Biosafety Cabinet
200 mm
0.99995
Energy efficient
2 x Each Side plus Smart Ports
50/60 Hz
230 V
Stainless Steel (316)
Herasafe 2030i
7 in. GUI
H14 HEPA
LED
7.8 in.
800
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.