Learn More
OHAUS™ Guardian 7000 Hotplate-Stirrer
Durable hotplate stirrers with superior safety features and intelligent performance
Brand:  OHAUS™ 30500623
 View more versions of this product
Product Code. 16600492
  • ONLINE EXCLUSIVE PRICE
    7810.00 DKK
    7311.74 DKK / Each
    Save 498.26 DKK
    6% Off
Only null left
In Stock
Add to basket
This item is not returnable. View return policy
Description
Description
  • Protect your lab with patented SmartPresence™ & SmartLink™ technologies. These exclusive features automatically shut off the heater if no one is detected after a user set "time out" period has passed.
  • An industry-leading early detection system that uses two independent safety controls to continuously monitor the electronics and shut off heating before an overtemperature condition occurs.
  • Precise stirring from 60 to 1600 rpm is made possible by the design of a powerful motor, strong magnet and software controlled ramp rate, ensuring secure magnetic coupling for viscous applications.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Aluminum
2.812 kg
e-G71HSRDM
126.000 mm
5.00 in.
173.00 mm
6.80 in.
2.600 kg
230 V
230 V, 4.6 A, 50/60 Hz
10.50 in.
+ 5°C – 380°C
50/60 Hz
267.00 mm
5.700 lb
EU
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.