Learn More
Gilson™ PIPETMANTM Pipet
GREENER_CHOICE
PIPETMAN brings a new level of comfort into Gilson range with a redesigned piston mechanism that reduces the force needed to aspirate and dispense liquids. PIPETMAN maintains the trusted durability, accuracy, and reliability of the original PIPETMAN pipettes with comfort.
Brand:  Gilson™ F144059P
 View more versions of this product
Product Code. 12356132
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
This item is not returnable. View return policy
Description
Description
  • Body (handle) in PVDF minimizes the hand-warming effect during pipetting and is resistant against aggressive chemicals
  • Direct volume reading in decimal digits helps prevent mistakes in volume setting
  • Comes with classic metal or plastic tip ejector
  • Clip ejector fits securely into the body of the pipette and helps avoid slipping over time
  • Redesigned seal, lighter ejection springs and high quality piston grease offer as low as 2 Newtons of force to pipet
  • High quality piston grease
Model Specific Features
  • Models P2 and P10 incorporate dual position tip ejectors for use with D10 (short) and DL10 (long) tips
Order Info
Trade in program available. Please contact your Fisher Scientific sales representative for more details.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
±3, ±4, ±8μL
No
Blue
Reusable
Ergonomic
P1000
Pipetman
Non-sterile
100 to 1000 μL
D200, D300, DF100ST tip
1
318 x 108 x 38 mm
36.5 cm
30.5 cm
12 cm
.21 kg
Certified Quality
D200, D300, DF100ST Tip
Autoclavable
Manual
D-200, D-300, DF100ST Tip
PVDF, Stainless Steel
1
No
Pipette
Extremely low pipetting forces, lightweight design
Plastic Tip Ejector
3 Years
14.37 in.
12.01 in.
4.72 in.
.46 lb
Blue
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.