missing translation for 'onlineSavingsMsg'
Få mere at vide

Gibco™ Recovery™ Cell Culture Freezing Medium

Artikelnummer. 11560446
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
This item is not returnable. View return policy

Product Code. 11560446

Brand: Gibco™ 12648010

This item is not returnable. View return policy

Complete, ready-to-use freezing medium for cryopreservation of a wide variety of mammalian cells, including CHO-S, CHO-K1, HEK 293, Jurkat, and NIH 3T3

For many years, researchers have used a common recipe for making freezing medium: High-glucose Dulbecco's Modified Eagle Medium (DMEM) supplemented with 10% serum and 10% DMSO. Recovery Cell Culture Freezing Medium is an optimized version of this classic formulation, using Gibco Bovine Serum and Fetal Bovine Serum to provide improved cell recovery and viability after thawing.

  • Suitable for a wide variety of mammalian cells
  • Can be used to freeze most mammalian cells cultured in DMEM, DMEM/F-12, MEM, RPMI 1640, and Ham's F-12
  • Quality tested for pH, osmolality, sterility, and endotoxin
  • Each lot is performance tested using CHO-K1 cells

Cell Counting, Viability & Cryopreservation, Cell Culture, Mammalian Cell Culture, Gene Expression Analysis & Genotyping, In Vitro Transcription, PCR & Real-Time PCR

Order Info

Shipped on dry ice.

Compliance

Manufactured at a cGMP-compliant facility registered with the FDA as a medical device manufacturer and certified to ISO 13485 standards.

TRUSTED_SUSTAINABILITY

Specifications

Cell Type Mammalian Cells
Form Liquid
Product Type Freezing Medium
Serum Level Standard Serum Supplementation
Sterility Sterile-filtered
With Additives High Glucose, Phenol Red, DMSO (10%)
Without Additives No Phenol Red
Product Line Gibco, Recovery
Quantity 50 mL
Shipping Condition Dry Ice
Purity or Quality Grade Research Grade
Content And Storage Storage conditions: -5 to -20°C. Protect from light.
Shipping conditions: Frozen
Shelf life: 12 months from date of manufacture
Culture Type Mammalian Cell Culture
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Methyl-1-butanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 3-Methyl-1-butanol

For Research Use Only. Not for use in diagnostic procedures.

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt