Learn More
Agilent BioTek Gen5 Qualification Package
For Gen5 Data Analysis Software
Brand:  Agilent BioTek 5320502
Product Code. 15728498
  • 1240.00€ / Each
Please to purchase this item. Need a web account? Register with us today!
Alert:
Product qualification packages contain only documentation and procedures. Some of these procedures require the use of QC test solutions or other accessories, but these are not included with the product qualification packages.
This item is not returnable. View return policy
Description
Description
The Gen5 Qualification Package includes:
  • Gen5 IQ (Installation Qualification) and Checklist
  • Data Reduction Qualification Plan and Checklist
  • Export / Report Qualification Plan and Checklist
  • Gen5 protocols, experiments, and data input files referenced by test procedures included in Package
Procedures with checklists are provided in both PDF and Microsoft Word formats.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Validation Services
USB flash drive
Gen5 IQ (Installation Qualification) and Checklist; Data Reduction Qualification Plan and Checklist; Export / Report Qualification Plan and Checklist; Gen5 protocols, experiments, and data input files referenced by test procedures included in Package
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.