Learn More
Brady™ FreezerBondz™ Ultra-thin Cryogenic Polyester Labels
Label curved frozen lab surfaces including tubes, vials and cryocanes with the ultra-thin B-492 polyester label material. Print almost anywhere with a durable Brady™ handheld label maker.
Brand:  Brady™ 170777
 View more versions of this product
Product Code. 17980690
  • € 111.00 / Each
Only null left
In Stock
Add to basket
This item is not returnable. View return policy
Description
Description
  • Ideal for laboratory use on curved surfaces including test tubes, vials and straw pipettes
  • Withstands chemicals, abrasions and extreme temperatures, including storage in liquid nitrogen
  • Made with a permanent acrylic adhesive for tight bonding onto smooth curved surfaces such as glass and polypropylene
Specifications
Specifications
Not Autoclavable
Permanent Acrylic
Matte
Polyester
Label
Component and Equipment Labeling, Data and Telecommunications Labeling, Electrical Labeling, General Identification, Laboratory Labeling
1 Tape/Cartridge
25 ft. x 0.5 in.
0.5 in.
0.5 in.
RoHS Compliant to 2011/65/EU (Pass 2015/863)
White
Continuous
Thermal Transfer
Non-sterile
BMP41, BMP51, M511
-320°F to 230°F
25 ft.
1.3 cm
M4C-500-492
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.