Learn More
Brady™ FreezerBondz™ Ultra-thin Cryogenic Polyester Labels
Label curved frozen lab surfaces including tubes, vials and cryocanes with the ultra-thin B-492 polyester label material. Print almost anywhere with a durable Brady™ handheld label maker.
Brand:  Brady™ 170898
 View more versions of this product
Product Code. 17801729
  • € 73.90 / Pack of 200
Only null left
In Stock
Add to basket
This item is not returnable. View return policy
Description
Description
  • Ideal for laboratory use on curved surfaces including test tubes, vials and straw pipettes
  • Withstands chemicals, abrasions and extreme temperatures, including storage in liquid nitrogen
  • Made with a permanent acrylic adhesive for tight bonding onto smooth curved surfaces such as glass and polypropylene
Specifications
Specifications
Not Autoclavable
Permanent Acrylic
Matte
Polyester
Label
Laboratory labelling
-196°C
200
-320°F to 230°F
0.0032 in
41.28 mm (W) x 15.24 mm (H) x 9.53 mm (Dia)
1.625 in
0.6 in
15.24 mm
9.53 mm
2011/65/EU, 2015/863
White
Die-cut
Thermal Transfer
Non-sterile
BMP51;BMP53;M511;M510
200 Sets / Box
41.28 mm (W) x 15.24 mm (H) x 9.53 mm (Dia)
-196°C to +110°C
0.081 mm
0.375 in.
41.28 mm
0.5 in.
0.375 in
M5-95-492
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.