Learn More
Thermo Scientific™ FlipStrip™ Microtube Racks with Lid
GREENER_CHOICE
Product Code. 10646391 Shop All MBP Molecular BioProducts Products
Change view
Click to view available options
Color:
Blue
Fluorescent Green
Fluorescent Green, Fluorescent Orange, Fluorescent Pink, Purple
Fluorescent Orange
Fluorescent Pink
Green
Purple
Red
Red, Green, Yellow, or Blue
Yellow
Unit Size:
Each
Pack of 12
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
10646391 Fluorescent Pink Each
10358711 Red, Green, Yellow, or Blue Pack of 12
10778072 Fluorescent Green, Fluorescent Orange, Fluorescent Pink, Purple Pack of 12
11837233 Green Pack of 12
10556562 Blue Each
10065210 Purple Each
12368182 Yellow Pack of 12
11894402 Fluorescent Orange Pack of 12
10177000 Fluorescent Green Each
10095090 Red Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 10646391 Supplier Thermo Scientific™ Supplier No. 8668
Only null left
Add to basket
Edge
Add to basket
Hold 0.2mL strips and 1.5mL tubes on one side and and 0.5mL tubes on the other side of this microtube rack with lid, which is made from 100% recycled plastic.
Hold 0.2mL strips and 1.5mL tubes on one side of the Thermo Scientific™ FlipStrip Microtube Rack with Lid, and 0.5mL tubes on the other. The rack is made from 100% recycled plastic.
  • Holds 0.2mL strips and 1.5mL tubes on one side, 0.5mL tubes on other
  • Stable, low-profile design with an alphanumeric grid for easy tube identification
  • Lid protects samples and allows racks to be stacked
  • Several colors available
TRUSTED_SUSTAINABILITY
Specifications
Color Fluorescent Pink
Holds Microtubes (0.5 mL), Microtubes (1.5 mL), Microtubes (2 mL)
Material Recycled Plastic
Shape Rectangular
Stackable Yes
No. per Case 12 Racks
Product Type Flipstrip Rack