missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech™ FlexAble CoraLite™ 488 Antibody Labeling Kit for Rabbit IgG
Shop All Proteintech Products
Click to view available options
Quantity:
10 x 0.5 μg Reactions
200 x 0.5 μg Reactions
50 x 0.5 μg Reactions
Unit Size:
10 reactions
200 reactions
50 reactions
Description
FlexAble CoraLite™ 488 Antibody Labeling Kit for Rabbit IgG is a novel antibody labeling kit that uses an affinity linker to conjugate CoraLite™ 488 in any buffer condition to rabbit IgG primary antibodies from any supplier.
- One labeling reaction requires only 0.5 μg of antibody, regardless of the antibody concentration
- Label your rabbit IgG antibody in <10 min without any additional equipment
Specifications
Specifications
Kit Contents | 50 μL CoraLite™ 488 FlexLinker for Rabbit IgG, 100 μL FlexQuencher for Rabbit IgG, 500 μL FlexBuffer, Manual, Protocol Magnet |
Quantity | 50 x 0.5 μg Reactions |
Content And Storage | 1 Year at -20°C or 6 Months at +4°C, Avoid Light |
Excitation/Emission | 500 nm/520 nm |
Labeling Target | IgG |
Species | Rabbit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction