missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mettler Toledo™ FiveGo™ F2 pH/mV Meters
Shop All Mettler Toledo Products
Click to view available options
Type:
Meter with Field Kit
Meter with Food Kit
Meter with Standard Kit
pH/mV Meter
Unit Size:
1 unit
Description
Includes
LE438 IP67 sensor, electrode clip, wrist strap, installation QuickGuide, pH buffer starter pack
Different combinations of portable meters, sensors and accessories are available, which are ideally suited for specific applications or samples.
- Robust, IP67 waterproof ABS housing
- Large, well-structured 3.1 in. segmented LCD display for easy viewing
- Intuitive button arrangement and simple menu for easy operation
- Ergonomic design provide comfort to both big and small hands
- Foldable foot stand provides optimal viewing while situated on a lab bench
- Withstands wet and demanding environments
- Includes compact carrying case
- Three point calibration with four predefined buffer groups
- ATC and MTC temperature compensation
- Features Automatic and manual endpoint recording, Acoustic endpoint signal, Visual endpoint signal
- BNC, Cinch sensor inputs
- Ideal for mobile applications in the laboratory, at-line, or outdoors
- pH Range: 0.00 to 14.00
- pH Resolution: 0.01
- pH Accuracy: ±0.01mV
- mV Range: -1999 to 1999mV
- mV Resolution: 1mV
- mV Accuracy: ±1mV
- Temperature Range: 0° to +100°C (32° to 212°F)
- Temperature Resolution: 0.1°C
- Temperature Accuracy: ±0.5°C
- Memory Size: 200 measurements, current calibration
- Operating Hours: >200 hrs.
- Power Supply: 4 x 1.5V AAA LRO3 or 4 x 1.2V AAA NiMH HR03
- Dimensions: 18.8 x 7.7 x 3.3cm (97.4 x 3 x 1.3 in.)
- Weight: 0.26kg (0.5 lb.)
- F2-Meter only: Suitable for applications that require a specific sensor not included in a kit
- F2 Food Kit: Suitable for solid and semi-solid samples such as meat, fruit, cheese, and more
- F2 Field Kit: Suitable for wide range of general aqueous samples and for measurements on the go
- F2 Standard Kit: Suitable for wide range of general aqueous samples including emulsions and suspensions
Specifications
Specifications
| Includes | LE438 IP67 sensor, electrode clip, wrist strap, installation QuickGuide, pH buffer starter pack |
| Accuracy (Temperature) | ±0.5°C |
| Accuracy (mV) | ±1 |
| For Use With (Application) | For wide range of general aqueous samples including emulsions and suspensions |
| Measurement Modes | pH |
| Calibration | Linear |
| Range (pH) | 0 to 14 |
| Resolution (pH) | 0.01 |
| Range (mV) | -1999 to 1999 |
| Resolution (mV) | 1mV |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction