missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ FiveEasy™ F20 pH/mV Meters

Compact, benchtop meter provides high quality pH/mV measurements with simple click of a button

Brand:  Mettler Toledo™ 30266626

588.46 EUR valid until 2025-12-20
Use promo code "24121" to get your promotional price.


 View more versions of this product

Product Code. 15543360

  • ONLINE EXCLUSIVE PRICE
    € 830.66
    € 765.00 / Each
    Save € 65.66
    7% Off
View Available Promotions
Only null left
In Stock
Edge
Add to basket

Includes: LE438 sensor, installation QuickGuide, pH buffer starter pack

Description

Description

  • Compact, space saving design with ABS housing
  • Remove sensor holder after use and store on the side of the meter
  • Large, well-structured 4.3 in. segmented LCD display for easy viewing
  • Intuitive button arrangement and simple menu for easy operation
  • ATC and MTC temperature compensation
  • Three point (max.) linear calibration with four predefined buffer groups
  • BNC, Cinch, NTC 30kiloOhm sensor inputs
  • Features analog output, Automatic and manual endpoint recording, Acoustic endpoint signal, Visual endpoint signal
  • Ideal for wide range of laboratory applications
Specifications:
  • pH Range: 0.00 to 14.00
  • pH Resolution: 0.01
  • pH Accuracy: ±:0.01
  • mV Range: 2000 to 2000mv
  • mV Resolution: 1mV
  • mV Accuracy: ±1mV
  • Temperature Range: 0° to +100°C (32° to 212°F)
  • Temperature Resolution: 0.1°C
  • Temperature Accuracy: 0.5°C
  • Memory Size: current calibration
  • Power Supply: 100 to 240V, 50/60Hz, 12 VDC
  • Dimensions: 22.7 x 14.7 x 7cm (9 x 5.7 x 2.7 in.)
  • Weight: 0.63kg (1.4 lb.)
Applications:
  • F20 Meter only: Suited for applications that require a specific sensor not included in a kit
  • F20 Standard Kit: Suited for wide range of general aqueous samples
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FiveEasy F20-Std
0.5°C
±0.01
Linear
0.01
1mV
32°F to 212°F
LCD
BNC, Cinch, NTC 30kiloOhm sensors
0.63 kg
F20
Meter with Standard Kit
50/60 Hz
F20 pH meter, LE438 pH sensor, electrode holder, 1 buffer sachets each for pH 4.01, 7, 9.21, and 10.01, test certificate, pH measurement guide
±1
Wide range of general aqueous samples
0 to 14
2000 to 2000
0°C to 100°C
0.1°C
Current calibration
22.7 x 14.7 x 7 cm (9 x 5.7 x 2.7 in.)
1.4 lb.
ATC and MTC
120/240 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.