missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Traceable™ Refrigerator/Freezer Alarm Thermometers

Provides accurate reading when freezer/refrigerator doors are open with temperature-buffered, sensor-sealed in bottled glycol solution

Brand:  Fisherbrand™ 90008-46

Product Code. 11705853

  • ONLINE EXCLUSIVE PRICE
    € 134.00
    € 82.65 / Each
    Save € 51.35
    38% Off
Only null left
In Stock
Edge
Add to basket

Includes: Velcro™ and magnetic mounting strips, one AAA battery, and ABS plastic case

Description

Description

Provides accurate reading when freezer/refrigerator doors are open with temperature-buffered, sensor-sealed in bottled glycol solution. Fisher Scientific™ Traceable™ Refrigerator/Freezer Alarm Thermometer shows current, high and low temperatures and alerts user with visual and audible signals if temperature diverts from set points.

  • Monitors temperature in freezers, refrigerators, incubators, water baths and heating blocks
  • Triple display simultaneously shows current, high and low temperatures
  • Alarm (Cat. No. 06-664-11 only) set in 1° increments alerts user with visual and audible signals if temperature goes outside setpoints
  • 10 ft. long (3m) micro cable permits doors to close on it
  • Dimensions: 4.25L x 2.75W x 0.75 in.H (11 x 7 x 2cm)

Certifications

Individually serial-numbered Traceable Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™; indicates traceability to standards provided by NIST™.

Specifications

Specifications

ISO 17025, A2LA, NIST
Monitoring temperature in freezers, refrigerators, incubators, heating blocks
16 mm
2.5 in.
63.5 mm
0.1°
0.11 kg
Refrigerator/Freezer Thermometer
158°F
-58°F
-58°F to +158°F
Yes
±0.5°C
82.5 mm
2.5 x 3.25 x 0.63 in. (63.5 x 82.5 x 16 mm)
0.63 in.
Velcro™ and magnetic mounting strip, one AAA battery, ABS Plastic case
63.5 mm
Plastic
4 oz.
3.25 in.
Bottle Probe
70°C
-50°C
-50°C to +70°C
Monitoring Temperature in Freezer, Refrigerator, Incubator, Heating Block
Digital LCD
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.