missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Fetal Bovine Serum, Value

Product Code. 17479633
Click to view available options
Quantity:
50 mL
500 mL
10 x 50 mL
Format:
Bottle
One Shot
Heat Inactivated:
No
Yes
Gamma-irradiated:
No
Yes
Unit Size:
500mL
50mL
Pack of 10
This item is not returnable. View return policy
This item is not returnable. View return policy

Includes

Gibco A5256701

Gibco fetal bovine sera offer an excellent value for basic cell culture, specialty research, and specific assays.

Gibco fetal bovine sera offer an excellent value for basic cell culture, specialty research, and specific assays. It has earned the trust of researchers with consistent quality and award-winning support that helps meet research needs and budget requirements.

Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that help you meet your research needs and budget requirements.

Gibco serum delivers

  • ISO 13485-certified, processed in FDA-registered facilities
  • Triple filtered at 0.1 μm
  • Gibco bottle is easier to use in hood, reduces risk of contamination, and helps you perform cell culture more consistently
  • Peel-off sticker on label provides handy reference for lot number and expiration date recording in lab notebook
  • Sera category: Value
  • Sera application: cell culture with robust cell lines, good value for basic research
  • Testing: up to 30 quality tests
  • Endotoxin level: ≤20 EU/mL (levels routinely ≤10 EU/mL)
  • Hemoglobin level: ≤25 mg/dL
  • Origin: refer to CoA for specific origin

All products may not be available in all regions due to importation regulations.

US Department of Agriculture (USDA) safety tested for the absence of bluetongue and akabane from countries that are USDA-approved for importation into the US.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at <-10°C.
Description Gibco A5256701
Species Cattle/Bovine
Age Fetal
Country of Origin Refer to CoA for specific origin
Form Liquid
Format Bottle
Product Type Fetal Bovine Serum
Quantity 500 mL
Serum Treatment Standard (Sterile-filtered)
Sterility Sterile
Sterilization Method Triple-filtered, 0.1 μm
Processing Triple Filtered at 0.1 μm
Tested For Up to 30 quality tests including 9CFR virus testing* endotoxin, performance (*Please note: If manufactured in the UK, Full 9CFR except for Rabies and BlueTongue Virus which is tested via EMA)
Endotoxin Concentration ≤20 EU/mL
Green Features Less waste
Heat Inactivated No
Hemoglobin Concentration ≤25 mg/dL
Purity or Quality Grade Value
Shipping Condition Dry Ice
USDA Safety Testing Included USDA Safety Testing Performed for Mexican and Australian origins
Gamma-irradiated No
Show More Show Less

For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.