missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Combitips Advanced™ Biopur™ Pipette Tips
GREENER_CHOICE

Product Code. 12674597 Shop All Eppendorf Products
Change view
Click to view available options
Volume (Metric):
0.1 mL
0.2 mL
0.5 mL
1 mL
2.5 mL
5 mL
10 mL
25 mL
50 mL
Color Code:
Blue
Green
Light Blue
Light Gray
Orange
Red
Violet
White
Yellow
Unit Size:
Pack of 100
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Color Code unitSize
12674597 5 mL Blue Pack of 100
12614597 0.1 mL White Pack of 100
12634597 0.2 mL Light Blue Pack of 100
12654597 0.5 mL Violet Pack of 100
12644597 1 mL Yellow Pack of 100
12664597 2.5 mL Green Pack of 100
12684597 10 mL Orange Pack of 100
12694597 25 mL Red Pack of 100
12604607 50 mL Light Gray Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 12674597 Supplier Eppendorf™ Supplier No. 0030089669

Please to purchase this item. Need a web account? Register with us today!

Edge

Meet the increased requirements of a modern laboratory with these pipetter tips. Eppendorf™ Combitips Advanced™ Biopur™ Pipetter Tips make working with radioactive, aggressive or toxic materials safer. Hermetically sealed piston prevents aerosol contamination.

Meet the increased requirements of a modern laboratory with these pipetter tips. Eppendorf™ Combitips Advanced™ Biopur Pipetter Tips make working with radioactive, aggressive or toxic materials safer. Hermetically sealed piston prevents aerosol contamination.

Combitips advanced are the ideal components for all Multipettes™. The new individual color coding facilitates quick identification of the desired tip.

  • Slip agent free
  • Color-coded tip size recognition
  • Funnel geometry from cylinder to rim, prevents damage to gloves and ensures comfortable handling
  • Function according to the positive displacement principle
  • Elongated design for complete emptying of all common tubes
  • Optimized graduations for visibility of liquid volume
  • Automatic Combitip recognition and volume calculation
  • 3D design on the upper rim for easy recognition
  • Maximum range offered by nine volume sizes (0.1 to 50 mL)
  • Dust free storage
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 5 mL
Sterility Sterile
Color Colorless
Autoclavable Not Autoclavable
Clean Claims Biopur™ (Sterile)
Format Individually Wrapped
Tip Style Extended Length, Positive Displacement
Packaging 100 Pk., Individually Blister-wrapped
Product Type Pipette Tips
Color Code Blue
Disposable Single-use
For Use With (Equipment) Eppendorf Multipette
Graduated Yes
Product Line CombiTips Advanced
Material Polypropylene
Fit for Purpose Sterile and PCR Ready
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.