Learn More
Eppendorf™ Combitips Advanced™ Biopur™ Pipette Tips
GREENER_CHOICE
Meet the increased requirements of a modern laboratory with these pipetter tips. Eppendorf™ Combitips Advanced™ Biopur™ Pipetter Tips make working with radioactive, aggressive or toxic materials safer. Hermetically sealed piston prevents aerosol contamination.
Brand:  Eppendorf™ 0030089650
1449.78 DKK valid until 2025-08-22
Use promo code "24148" to get your promotional price.
 View more versions of this product
Product Code. 12664597
  • 1526.08 DKK / Pack of 100
Focus on Agri-Science Testing
Use promo code 24136 at checkout
View Available Promotions
Only null left
In Stock
Edge
Add to basket
Description
Description
Meet the increased requirements of a modern laboratory with these pipetter tips. Eppendorf™ Combitips Advanced™ Biopur Pipetter Tips make working with radioactive, aggressive or toxic materials safer. Hermetically sealed piston prevents aerosol contamination.
Combitips advanced are the ideal components for all Multipettes™. The new individual color coding facilitates quick identification of the desired tip.
  • Slip agent free
  • Color-coded tip size recognition
  • Funnel geometry from cylinder to rim, prevents damage to gloves and ensures comfortable handling
  • Function according to the positive displacement principle
  • Elongated design for complete emptying of all common tubes
  • Optimized graduations for visibility of liquid volume
  • Automatic Combitip recognition and volume calculation
  • 3D design on the upper rim for easy recognition
  • Maximum range offered by nine volume sizes (0.1 to 50 mL)
  • Dust free storage
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2.5 mL
Colorless
Biopur™ (Sterile)
Extended Length, Positive Displacement
Pipette Tips
Single-use
Yes
Polypropylene
Sterile
Not Autoclavable
Individually Wrapped
100 Pk., Individually Blister-wrapped
Green
Eppendorf Multipette
CombiTips Advanced
Sterile and PCR Ready
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.