missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck MultiScreen™HTS 8-Well Strips with Immobilon™-P PVDF Membrane
Shop All Merck Products
Click to view available options
Product Type:
Microplate 8-Well Strip
Microplate Strip Support Frame
Unit Size:
Pack of 10
Description
Includes
Twelve 8-well strips in a support frame. Additional support frames also available separately.
- For ELISpot and protein binding assays
Specifications
Specifications
| Format | Strip Plate |
| Array | 1 x 8 |
| Color | Clear |
| Product Type | Microplate 8-Well Strip |
| Material | ABS Plastic |
| No. of Wells | 8 |
| Sterility | Sterile |
| For Use With (Application) | ELIspot and protein binding assays |
| Quantity | 10/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction