missing translation for 'onlineSavingsMsg'
Learn More

Merck MultiScreen™ 96-Well Assay Plates for ELISPOT Assays

Product Code. 10146580 Shop All Merck Products
Click to view available options
Color:
Clear
White
Material:
Acrylic
Polystyrene
Sterility:
Non-sterile
Sterile
Membrane:
Hydrophobic PVDF
Hydrophobic PVDF with Underdrain
Mixed Cellulose Esters
Unit Size:
Pack of 10
Pack of 50
This item is not returnable. View return policy
This item is not returnable. View return policy

Give optimal results in immunological ELISPOT (enzyme-linked immunospot) assays by providing high protein-binding capacity with low background staining

  • Membrane-to-plate seal ensures no reagent crosstalk and limited optical crosstalk Patented underdrain support structure allows liquid volumes to be maintained for incubation times of up to 21 days

TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Lid Yes
Membrane Hydrophobic PVDF with Underdrain
Array 8 x 12
Pore Size 0.45 μm
Product Line MultiScreen
Color Clear
Product Type MultiScreen-IP with Underdrain
Material Acrylic
No. of Wells 96
Sterility Sterile
Filtration Area 0.28 cm2
For Use With (Application) ELISPOT Assays
Quantity 10/Pk.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.