Learn More
Merck Millicell™ Receiver Trays
Product Code. 10052523 Shop All Merck Products
Change view
Click to view available options
Well Count:
1
24
Unit Size:
Each
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Well Count unitSize
10052523 24 Each
10347753 1 Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10052523 Supplier Merck Supplier No. PSMW010R5
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Comes with lid
Designed to support cell attachment, growth, and differentiation for a variety of applications
  • 24-well feeder tray incorporates a tear-drop well design to reduce the incidence of air bubbles that can form under the membrane during plate assembly, ensuring that cell monolayers are getting the proper nutrients
  • Single-well feeder tray utilizes baffles to reduce media leakage and contamination
TRUSTED_SUSTAINABILITY
Specifications
Closure Included With Lid
Color Clear
Material Polystyrene
Shape Rectangular
Sterility Sterile
Type Receiver Tray
Well Count 24
Includes 24-Well Receiver Tray, Lid
Supportive Membrane PCF