missing translation for 'onlineSavingsMsg'
Learn More

Sel disodique EDTA en solution aqueuse, BAKER ANALYZED™, J.T. Baker™

Code produit 15548424 Tous les produits J.T.Baker Produits
Change view
Click to view available options
:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15548424 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15548424 Supplier JT Baker Supplier No. 7613.1000

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

This item is not returnable. View return policy

Réactif de laboratoire

  • Molarité : 0,0099 - 0,0101

Specifications

CAS 139-33-3
Conditionnement Flacon en plastique
Formule moléculaire C10H14N2Na2O8
Numéro MDL MFCD00070672,MFCD00003541,MFCD00070672,MFCD00150037
Quantité 1 L
Synonyme Edetic acid disodium salt, Disodium dihydrogen ethylenediaminetetraacetate, Ethylenediaminetetra-acetic acid disodium salt, Idranal III, Kestranal™ 2 S, Ethylenedinitrilotetra-acetic acid disodium salt
Clé InChI ZGTMUACCHSMWAC-UHFFFAOYSA-L
SMILES [Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
Nom IUPAC acétate disodique de 2-({2-[(carboxylatométhyl)(carboxyméthyl)amino]éthyl}(carboxyméthyl)amino)
Poids moléculaire (g/mol) 336.21
Concentration or Composition (by Analyte or Components) 0,01 mol/l (0,02 N)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.