missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Plain Economy PTFE Stir Bar

Product Code. 11597572 Shop All Fisherbrand Products
Change view
Click to view available options
Length (Metric):
12 mm
25 mm
40 mm
50 mm
Diameter (Metric):
3 mm
8 mm
Unit Size:
Each
Pack of 10
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) unitSize
11597572 25 mm 8 mm Pack of 10
11517582 50 mm 8 mm Pack of 10
11587572 12 mm 3 mm Pack of 10
11507582 40 mm 8 mm Pack of 10
10373692 40 mm 8 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11597572
Only null left
Add to basket
Add to basket

Provides stable stirring action. The plain economy PTFE magnetic stir bar work within a wide range of conditions, low prices from high-volume manufacturer.

  • Stable, general-purpose magnetic stir bar
  • Smooth, round profile for a wide variety of applications
  • Autoclavable

Specifications

Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Color White
Description PTFE encapsulated Alnico V magnet core
For Use With (Equipment) Containers & Vessels / Stirrer Hot Plates
Diameter (Metric) 8 mm
Length (Metric) 25 mm
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.