Learn More
DWK Life Sciences DURAN™ PURE Premium Screw Cap
BIOPHARMA
Product Code. 11679808 Shop All DWK Life Sciences Products
Change view
Click to view available options
Thread Size (Metric):
GL 25
GL 45
Unit Size:
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thread Size (Metric) unitSize
11679808 GL 45 Pack of 5
12866573 GL 25 Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11679808 Supplier DWK Life Sciences Supplier No. 891088679
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
High-quality closures developed for the needs of the pharmaceutical and biotech industry
High-quality plastic screw caps with GL 25/GL 45 thread sizes, developed for the needs of the pharmaceutical industry. DURAN™ PURE Premium Screw Caps are manufactured in clean rooms and made from PFA with highly inert PTFE coated silicone seal. Designed for use with DURAN™ PURE GL 25/ GL 45 Bottles. The screw caps are compatible with autoclaving and other common sterilization methods. DWK Life Sciences provides CoAs with each delivery, assures 100% traceability and change control management. DURAN™ PURE Premium Screw Caps are available with the documentation required by regulatory authorities and meet the requirements of the international pharmacopoeias.
  • High cleanliness due to manufacturing in clean rooms
  • Single batch delivery
  • Certificate of Analysis (CoA)
  • Visual and dimensional inspection
  • Change control management and information of customer in case of changes
  • 100% traceability of all manufacturing steps
  • Extractables report available
  • Drug Master File
  • USP <88> Class VI Pharmaceutical grade materials
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
For Use With (Application) Manufacturing of pharmaceutical drugs and biotech products; Storage and transport of APIs, bulk drug intermediates, ingredients and media
Material PFA, PTFE, Silicone
Quantity 5/Pk.
Thread Size (Metric) GL 45
Diameter (Metric) 51 mm
Autoclavable Autoclavable