missing translation for 'onlineSavingsMsg'
Learn More

Welch Ilmvac™ Diaphragm Vacuum Pump Service and Maintenance Kit

Product Code. 10090972 Shop All Welch Ilmvac Products
Change view
Click to view available options
For Use With:
MP/MPC 101 Z, 201 E
MP/MPC 201 T, MP 101V
MP/MPC 301 Z, 601 E, 301 Z
MP/MPC 601 T, 901 Z, 1201 E, MP 301V
MP/MPR 060 E, 030 Z, Biovac 060, MP 101V
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With unitSize
10090972 MP/MPC 601 T, 901 Z, 1201 E, MP 301V Each
13191984 MP/MPC 101 Z, 201 E Each
10402742 MP/MPC 201 T, MP 101V Each
10245242 MP/MPC 301 Z, 601 E, 301 Z Each
10328687 MP/MPR 060 E, 030 Z, Biovac 060, MP 101V Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10090972 Supplier Welch Ilmvac™ Supplier No. 402042
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Service and Maintenance Kits comprised of replacement seals and wearable parts for various models of diaphragm pumps.

  • Available for a range of diaphragm vacuum pump models
  • Provided individually
TRUSTED_SUSTAINABILITY

Specifications

For Use With MP/MPC 601 T, 901 Z, 1201 E, MP 301V
Includes Diaphragm, Valves, and O-rings
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.