missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Grant Instruments™ Dry Block Heating System, BTD Series
BTD compact and flexible fixed block system for rapid and precise heating of microtubes up to 100ºC.
Marca: Grant Instruments™ BTD
5149.76 DKK valid until 2025-12-20
Use promo code "24121" to get your promotional price.
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The Grant BTD holds a combination of four microtube sizes simultaneously, and rapid heat-up enables swift change of application for maximum flexibility. The aluminum block is housed in a compact, stylish plastic moulding, which is durable, sturdy and easy to clean.- Digital temperature control for optimum precision
- Heating range ambient + 5°C to 100°C
- Powerful heater for rapid heat-up times - 25°C to 100°C in just 15 minutes, 25°C to 37°C in just 4 minutes
- Stability ± 0.1°C, uniformity ± 0.2°C
- Heating block holds combinations of four microtube sizes simultaneously, up to a total of 49 tubes: 24 x 1.5/2.0mL, 15 x 0.5mL, 10 x 0.2mL
- Flexible tube sizes and rapid heat-up time enables swift change of application
- Convenient integral timer for time-sensitive incubations
- Sturdy, durable, easy-to-clean plastic construction; compact design with small footprint
- Two-line display for simple and precise setting of temperature/time and for monitoring current status during operation

Especificaciones
Ambient +5° to 100°C | |
Life-science/cancer research - DNA extraction incubations, DNA denaturation, PCR, ELISA and Western blotting, molecular biology | |
±0.1°C | |
230 x 210 x 110 mm | |
115 mm | |
120 or 230 V | |
50/60 Hz |
Dry Bath Block | |
0.1°C | |
1 minute to 96 hours (increments of 1 minute) | |
230 mm | |
2.8 kg | |
200 w |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido