missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ Double Bagged - USP Class VI Liquid Funnels
BIOPHARMA

Product Code. 17584706 Shop All Sampling Systems Products
Change view
Click to view available options
Diameter (Metric)Top:
50 mm
100 mm
Unit Size:
Pack of 20
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric)Top unitSize
17584706 100 mm Pack of 20
17594706 50 mm Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17584706 Supplier Sampling Systems™ Supplier No. 9370H100S
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Our premium range of Liquid Funnels are made from high grade medical USP Class VI HDPE. It is ideal for demanding applications such as vaccine manufacture, GMP, and cleanroom manufacturing.

  • The SteriWare™ Ultra range of liquid funnels are available in both 50 mm and 100 mm diameter.
  • This premium funnel range is double bagged.
  • A box liner is also used giving a third layer of protection.
  • It is ideal for demanding applications such as vaccine manufacture, GMP and cleanroom manufacturing.
  • The funnels all have an ergonomic lug on the side so handling them cleanly is easy.
  • The funnels are designed to be used straight from the bag.
  • Made from food grade, FDA and EC compliant HDPE.
  • HDPE has good chemical resistance and can be easily recycled.
  • The funnels are sterilised by gamma irradiation.
  • USP Class VI - High quality medical grade polymer that has also been independently tested to USP 661.
  • Cleanroom Manufactured - The funnels are entirely made in cleanroom conditions with final packing carried out in a class 10,000 cleanroom.
  • Each funnel is 100% inspected before packing.

Specifications

Diameter (Metric)Top 100 mm
Material HDPE
Sterility Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.