missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DNA AWAY™ Surface Decontaminant

Product Code. 10223471 Shop All MBP Molecular BioProducts Products
Change view
Click to view available options
Description:
DNA Surface Decontaminant
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description unitSize
10223471 DNA Surface Decontaminant 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
Product Code. 10223471 Supplier Thermo Scientific™ Supplier No. 7010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price

Eliminate unwanted DNA and DNase from glassware and plasticware without affecting subsequent DNA samples. This surface decontaminant degrades DNA more quickly and effectively than autoclaving.

Eliminate unwanted DNA and DNase from glassware and plastic ware without affecting subsequent DNA samples. Thermo Scientific™ DNA AWAY™ Surface Decontaminant degrades DNA more quickly and effectively than autoclaving.

  • Suitable for gel boxes, pipettes, benchtops, thermal cyclers, or other apparatus
  • Use before performing PCR or working with DNA for a DNA-free work area
  • Simply apply the ready-to-use formula to the surface to decontaminate, then wipe dry or rinse clean
TRUSTED_SUSTAINABILITY

Specifications

pH 11
Composition Sodium Hydroxide with Water
Description DNA Surface Decontaminant
Volume (English) 8.5 oz.
Volume (Metric) 250 mL
Supplemental information
  • MIXTURE LIST-Contain: sodium hydroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.