missing translation for 'onlineSavingsMsg'
Learn More

Grant Instruments™ DEN-600 - Densitometer, Portable Battery/Mains, Cell Photometer Turbidity 0 - 16 McF & 0.3 Abs

Product Code. 17541136
Click to view available options
Type:
Photometer
Unit Size:
Each
This item is not returnable. View return policy
This item is not returnable. View return policy

The DEN-600 is a compact, portable, rechargeable battery powered photometer. It’s 600 nm wavelength optical system allows you to measure estimated total number of cells (OD600), turbidity measurement method (McFarland (McF)), protein concentration measurement (Bradford protein assay method)

The device serves as an inexpensive alternative to a spectrophotometer, which is commonly used for these applications. As the DEN-600 is battery powered and compact, it can be comfortably located in a biosafety cabinet, anaerobic chamber or quickly relocated.

The cell holder accommodates standard 10 mm path cuvettes, round bottom, conical vials or falcon tubes.

USB connectivity and DEN software allow for data transfer, data processing and calculation.

Applications

  • Cell concentration measurement
  • Cell growth data estimation
  • Log phase estimation for microbial cells induction
  • Competent cell preparation
  • Bradford protein assay method
  • Antibiotic susceptibility testing
  • Inhibitory tests
TRUSTED_SUSTAINABILITY

Specifications

Type Photometer
Display Type LED
Accuracy ±0.006 at 1 Abs, ±0.1 at 0 to 8 McF
Depth (Metric) 145 mm
For Use With (Equipment) For 10 mm path cuvettes, round bottom and falcon tubes
Voltage 120/240 V
Height (Metric) 65 mm
Light Source LED
Wavelength 600 nm
Weight (Metric) 0.5 kg
Width (Metric) 120 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.