missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IBA Lifesciences Desthiobiotin
Shop All IBA Lifesciences Products

Click to view available options
Quantity:
1 g
5 g
Unit Size:
Each
Description
Desthiobiotin is a modified form of biotin that displaces Strep-tag™II and Twin-Strep-tag™ fusion proteins from the modified biotin-binding pocket of Strep-Tactin™ in a competitive manner, resulting in a mild elution.
Desthiobiotin powder can be used for preparation of Strep-Tactin™ elution buffer, Buffer E.
Please note that desthiobiotin is not applicable for elution from Strep-Tactin™XT resins.
Specifications
Specifications
| Chemical Name or Material | Desthiobiotin |
| For Use With (Application) | For Elution of Strep-Tag II Proteins From Strep-Tactin |
| Quantity | 1 g |
For Research Use Only.
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion