Learn More
3M™ Safety-Walk™ Cushion Matting 5270E
This motor oil resistant mat, suitable for heavy foot traffic in dry areas, provides a comfortable surface for walking or standing.
€ 475.00 - € 4785.00
Specifications
Color Black
Type Cushion Mat
View More Specs
Products 3
Product Code Brand Length (Metric) Price Quantity & Availability  
Product Code Brand Length (Metric) Price Quantity & Availability  
17148740
View Documents
3M™ Safety-Walk™
7100116724
3.1 m This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
17138740
View Documents
3M™ Safety-Walk™
7100116723
6.1 m This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
17128740
View Documents
3M™ Safety-Walk™
7100116722
9.1 m This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
  • Coiled cushion matting compresses evenly across the bottom of the shoe, reducing leg and back fatigue
  • Textured top improves traction for safer footing
  • Suitable for dry work areas such as assembly lines and production areas
  • Supplied with edging to allow for wheeled traffic
  • Withstands most industrial and cleaning chemicals
Specifications
Specifications
Black
Cushion Mat
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.