Learn More
Thermo Scientific™ CultiMaxx™ Shelving Systems for Nunc™ Cell Factory™ System
CultiMaxx™ Shelving System to support Nunc™ Cell Factory™ systems
Brand:  Thermo Scientific™ 50166285
 View more versions of this product
Product Code. 17851700
  • 2130.60 € / Each
Only null left
In Stock
Add to basket
This item is not returnable. View return policy
Description
Description
These shelving systems are designed to support scale-out of the Nunc™ Cell Factory™ System in cell therapy production. With an in-depth review of the shelving design, this new system provides less than 0.5mm of deflection per vessel, keeping the media level to promote even cell growth conditions. Individual access to each vessel allows for minimal disruption to adherent cells when removing samples from the incubator. The staggered shelving further enables flexibility by providing the necessary clearance to leave filters and connections in place when loading the systems into the incubator.
These shelving systems are designed to support scale-out of the Nunc Cell Factory System in cell therapy production. With an in-depth review of the shelving design, this new system provides less than 0.5 mm of deflection per vessel, keeping the media level to promote even cell growth conditions. Individual access to each vessel allows for minimal disruption to adherent cells when removing samples from the incubator. The staggered shelving further enables flexibility by providing the necessary clearance to leave filters and connections in place when loading the systems into the incubator.
Easy Installation
  • Shelving systems are retrofittable and can be self-installed.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Shelving system to support up to 6 Nunc™ Cell Factory™ system with 10 or 13 layers for Heracell™ VIOS™ 250i (CR), Forma™ Steri-Cycle™ i250 (CR) CO2 Incubator
Stainless Steel
Accessory
Heracell™ Vios™ 250i, Forma™ Steri-Cycle™ i250, Heracell™ Vios™ 250i CR, Forma™ Steri-Cycle™ i250 CR CO2 Incubators, Heracell Vios 250i, Forma Steri-Cycle i250 and CR/CTS series
3
3 shelves with reinforced design and staggered to allow filter clearance, shelf supports and shelf rails
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.