Learn More
Thermo Scientific™ Culti-Loops™ Pseudomonas aeruginosa ATCC™ 27853™
Beschreibung
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
Spezifikation
Spezifikation
| ATCC® | 27853TM* |
| Description | Pseudomonas aeruginosa ATCC™ 27853™ |
| Format | Culti-Loops |
| Product Type | Culti-Loop |
| Target Organism Class | Gram-negative Rods |
| Quantity | 5 Loops/Pack |
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.