Learn More
Corning™ HYPERFlask™ M Cell Culture Vessels
BIOPHARMAGREENER_CHOICE
Product Code. 10281845 Shop All Corning Products
Change view
Click to view available options
Surface Coating:
CellBIND Treated
Not Treated
No. per Pack:
1/Pk.
4/Pk.
No. per Case:
24/Cs.
4/Cs.
Unit Size:
Pack of 24
Pack of 4
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Surface Coating No. per Pack No. per Case unitSize
10281845 CellBIND Treated 1/Pk. 4/Cs. Pack of 4
10569765 CellBIND Treated 4/Pk. 24/Cs. Pack of 24
10343305 CellBIND Treated 4/Pk. 4/Cs. Pack of 4
15554449 Not Treated 1/Pk. 4/Cs. Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10281845 Supplier Corning™ Supplier No. 10030
Only null left
Add to basket
Add to basket
Designed specifically for manual use, high-yield and high-performance. Specifically designed for manual use. Corning™ HYPERFlask™ M Cell Culture Vessel uses a multi-layer gas permeable growing surface for efficient gas exchange.
The Corning High Yielding Performance Flask (HYPERFlask) cell culture vessel offers 1720 cm² growth area in the footprint of a traditional 175 cm² flask. This high yield, high performance flask utilizes a multilayered gas permeable growing surface for efficient gas exchange.
  • Innovative Design – 10 interconnected polystyrene growth surfaces
  • Optimal Growth – Corning CellBIND™ Surface treated gas permeable polystyrene for superior cell attachment and growth
  • Increase Cell Yield – 10-fold higher cell yields increases productivity and capacity and reduces processing time and incubator space
  • Save Time and Space – Reduce processing time and incubator storage space by handling one flask as compared to 10 traditional 175 cm² flasks
  • Automation Compatible – Bar coded and compatible
  • with The Automation Partnership SelecT™ and CompacT™ SelecT™ cell culture systems with an upgrade
The Corning HYPERFlask Cell Culture Vessel has the same external dimensions as a standard T 175 cm² flask with 10 times the cell yield.
Order Info
Supplied double bagged.
TRUSTED_SUSTAINABILITY
Specifications
Barcode Yes
Closure Color Orange
Closure Material HDPE
Capacity (Metric) 560 mL
Material Polystyrene
Neck Style Straight
No. per Case 4/Cs.
No. per Pack 1/Pk.
Shape Rectangular
Surface Coating CellBIND Treated
Graduated No
Sterility Sterile
Bottom Shape Rectangular
Closure Type Flat