Learn More
Proteintech CoraLite™488-Annexin V and PI Apoptosis Kit
CoraLite™488-Annexin V and PI Apoptosis Detection Kit provides a quick and easy method to detect apoptosis by marking early apoptotic cells (green) and late apoptotic cells or necrotic cells (red).
Brand:  Proteintech PF00005-100T
Product Code. 17368252
  • 1305.99 DKK / 100 tests
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
  • Results can be read out using flow cytometry or other fluorescence testing equipment.
  • Early apoptic cells are identified by annexin V (labeled with CoraLite(R) 488 dye) binding to phosphatidylserine (PS) flipped to the extracellular membrane surface, an early event in apoptosis.
  • Compared with FITC, CoraLite(R) 488 dye has higher fluorescence brightness, and is not affected by pH in the environment, and has good light stability.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates