missing translation for 'onlineSavingsMsg'
Learn More

Contec™ PROSAT™ Sterile Presaturated Nonwoven Wipes
BIOPHARMAGREENER_CHOICE

Product Code. 17126474 Shop All Contec Products
Change view
Click to view available options
:
Polypropylene; 9 x 11 in.; 70% denatured ethyl alochol and 30% deionized water
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17126474 Polypropylene; 9 x 11 in.; 70% denatured ethyl alochol and 30% deionized water
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17126474 Supplier Contec™ Supplier No. PS911EB/ETOH

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Wipe down isolators, cabinets, hoods, and other cleanroom equipment and tools. Contec™ PROSAT™ Sterile Presaturated Nonwoven Wipes are ideal for cleanup following BD RODAC™ plate sampling. These wipes are designed specifically for sterile environments.

  • Designed for pharmaceutical, biotech, and medical device applications
  • Choose spunlaced nonwoven 100% polyester wipes for improved cleanliness, economical hydroentangled polyester/cellulose wipers, or meltblown polypropylene wipes for improved durability
  • Conveniently presaturated with 70/30 blend of semi-conductor grade IPA and DI water or 70/30 blend of denaturated ethyl alcohol and DI water to reduce VOC emissions and hazardous waste
  • All wipes are sterilized by gamma radiation to a 10-6 Sterility Assurance Level and validated per AAMI ISO 11137 (Method 1) guidelines
  • Certificate of Sterility and Certificate of Irradiation included
  • Recommended for use in ISO Class 5 (FED-STD-209E Class 100/M3.5) environments
  • Individually lot numbered resealable pouches with tamper-evident labels ensure product integrity and cleanliness
TRUSTED_SUSTAINABILITY

Specifications

Product Type PROSAT™ Pre-saturated Wipes
Length (English) 11 in.
Length (Metric) 27.9 cm
Width (English) 11 in.
Width (Metric) 23 cm
Material Polypropylene
Certifications/Compliance ISO Class 4
Dimensions (L x W) 11 x 9 in. (28 x 23 cm)
For Use With (Application) Ideal for routine cleaning of tools, instruments and other laboratory equipment; designed for sterile environments
Packaging Quantity 30 Wipes/Bag
Saturation Solution 70% denatured ethyl alcohol/30% deionized water
Sterility Sterile
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • CONTEC PROSAT STERILE MBPP WITH 70% DENATURED ETHANOL
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P370+P378-In case of fire: Use dry extinguishing powder to extinguish.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P411-Store at temperatures not exceeding °C/°F.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: methanol ethanol, propan-2-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.