Learn More
Contec™ HydroPure

Blend of 6% hydrogen peroxide and purified water, low residue, sporicidal biocide. HydroPure is filtered to 0.2μ leaves no residue, eliminating any residue removal stage.
Brand: Contec™ FBT16HP
679.28 DKK valid until 2025-09-30
Use promo code "27072" to get your promotional price.
Description
Blend of 6% hydrogen peroxide and purified water, low residue, sporicidal biocide. Contec™ HydroPure is filtered to 0.2μ leaves no residue, eliminating any residue removal stage.
- Efficacious against bacteria, fungi, moulds, yeasts and spores
- 0.2μm filtered and filled under Grade A uni-directional flow air and bagged in Grade B cleanroom
- Leaves little to no residue and has endotoxin level of <0.25 EU/mL
- Safe to use in product contact areas
- Designed for ease of entry into pharmaceutical cleanrooms
- Ensures the product is free from contamination and particulates
- Sufficient sporicidal activity for Grade A/B area
- Large droplet size reduces the risk of inhalation and provides good surface coverage
- Bagged in polyethylene linear tear bags
- Facilitates transfer disinfection into isolators and RABS
- RTU with one year shelf life

Specifications
Hydrogen Peroxide and Purified Water Blend | |
Not Applicable | |
Non-sterile | |
Not Autoclavable | |
6% hydrogen peroxide in purified water | |
Limited-use | |
Yes | |
1 L |
Liquid | |
Unscented | |
Clear | |
Clear | |
Trigger Spray Bottle | |
Filtered | |
6 x 1 L |
Safety and Handling
- contec hydropure
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.