missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mettler Toledo™ Conductivity Meter SevenDirect SD30
Shop All Mettler Toledo Products

Click to view available options
Type:
SevenDirect SD30 Kit
SevenDirect SD30 Pure H2O Kit
SevenDirect SD30, Meter Only
Unit Size:
Each
Description
SevenDirect instruments are benchtop meters developed for a wide range of laboratory applications, which provide accurate measurements of conductivity of almost any sample. The SevenDirect is the successor of the SevenCompact and a great addition to any lab.
- Large color touchscreen provides direct measurement and easy data entry.
- Instructions guide users through the sensor calibration process.
- View detailed measurement information on one measurement screen - or focus on the result only and maximize readability with the uFocus mode.
- Closed connector compartment and replaceable cover keep the instrument safe from dust and spills, even with sensors attached (IP54 in-use).
- The instrument stores data like timestamp, sample and sensor ID with every measurement and prints or exports records automatically.
- A two-level user management protects the most important settings.
- The EasyPlace electrode arm makes placing the sensor in the sample or buffer container easy.
- Dedicated positions for cleaning and storage support correct sensor handling.
Specifications
Specifications
| Accuracy (Conductivity) | 0.005 |
| Measurement Modes | Conductivity |
| Range (Conductivity) | 0.001 μS/cm to 2000 mS/cm |
| Resolution (Conductivity) | 0.001 to 1 |
| Range (Salinity) | 0 to 80 ppt |
| Range (TDS) | 0 g/L to 100 g/L |
| Temperature Range (Metric) | -30°C to +130°C |
| Temperature Range (English) | -22°F to +266°F |
| Resolution (Temperature) | 0.1°C |
| Accuracy (Temperature) | ±0.1°C between 0°C and 100°C |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction