missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Grant Instruments™ Combi-Spin PVC 2400 Centrifuge/Vortex Mixer
Cost-effective, fixed-speed, combined micro centrifuge/vortex mixer for combined or independent centrifuge and mixing applications of microtubes and 0.2ml microtube strips in low volume applications.
Brand: Grant Instruments™ PCV-2400
2775.94 DKK valid until 2025-12-20
Use promo code "24121" to get your promotional price.
This item is not returnable.
View return policy
Description
- Temperature setting range +4ºC to +40ºC
- Centrifugation speed: fixed at 2800rpm (50Hz) and 3500rpm (60Hz)
- Choice of interchangeable rotors for different microtube sizes/combinations and for 0.2mL strips

Specifications
2800 rpm at 50 Hz, 3500 rpm at 60 Hz | |
1.5/2 mL, 0.5 mL, 0.2 mL | |
12 x 1.5 to 2.0 mL/12 x 0.5 mL plus 12 x 0.2 mL |
120/230 V | |
190 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction