Learn More
Memmert™ CO2 Incubator ICOmed (Medical Device)
Ideal for the creation and maintenance of constant environmental conditions for application in the field of in vitro fertilisation (IVF), especially for the incubation of oocytes, spermatozoa and zygotes in special culture dishes for IVF application.
€ 5702.00 - € 10420.00
Specifications
Alarm Types | Optic and acoustic |
---|---|
Capacity (Metric) Shelf | 15 kg |
Certifications/Compliance | DIN 12880:2007-05 , EN 61010-1 (IEC 61010-1), EN 61010-2-010 |
Controller Type | TwinDISPLAY: adaptive multifunctional digital PID-microprocessor controller with 2 high-definition TFT-colour displays |
Chamber Material | Stainless Steel |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
15799639
|
Memmert™
ICO50med |
56 L |
€ 6275.00 € 5702.00 / Each Save € 573.00 9% Off |
Estimated Shipment: 30-12-2025
Log in to see stock. |
|||||
15841575
|
Memmert™
ICO105med |
107 L |
€ 7505.00 € 6819.00 / Each Save € 686.00 9% Off |
Estimated Shipment: 04-11-2025
Log in to see stock. |
|||||
15851575
|
Memmert™
ICO150med |
156 L |
€ 9380.00 € 8527.00 / Each Save € 853.00 9% Off |
Estimated Shipment: 03-12-2025
Log in to see stock. |
|||||
15861575
|
Memmert™
ICO240med |
241 L |
€ 11465.00 € 10420.00 / Each Save € 1045.00 9% Off |
Estimated Shipment: 30-12-2025
Log in to see stock. |
|||||
Description
Sterile, safe and reliable – these three words describe the Memmert CO2 incubator ICOmed for the medical use.
Memmert subjected its CO2 incubator ICOmed to a comprehensive medical device evaluation. Each Memmert CO2 incubator ICOmed is classified as a Class IIa medical device for the in vitro fertilisation (IVF) as well as biosynthesis.
- Model sizes: 50, 105, 150, 240
- Temperature adjustment range: +18 °C to +50 °C
- CO2 adjustment range: 0 to 20 %
- Humidity adjustment ranges: 40 to 97 % rh (with option K7)
- O2 adjustment ranges: 1 to 20 % (with option T6)
- Classified as a medical device class IIa
- 1 model variant: TwinDISPLAY (intuitive control)
- Perfect distribution of temperature, humidity and CO2
- Long-time stable and repetitively accurate
- The interior chamber, including all installations and sensors, can be sterilised at +180 °C in a 60-minute programme
- Visual and acoustic alarm as well as alarm notification to mobile phones if the individually adjustable temperature, CO2, O2, or humidity ranges are exceeded
- Battery-buffered ControlCOCKPIT: The operating display, logging and CO2 control are fully functional even during a power failure
- Fail-safe, FDA-compliant logging of temperature, CO2, O2 humidity, open door time data
- Adjustable parameters: temperature (Celsius or Fahrenheit), CO2, programme time, time zones, summertime/wintertime
Active humidity control ensures short recovery times after the door has been opened and minimises vaporisation in the interior. Together with the heating of the interior from all six sides and the heated inner glass door, it offers maximum protection for cell and tissue cultures and avoids the dangerous formation of condensation. The turbulence-free chamber ventilation ensures a constant and uniform atmosphere.
Specifications
Optic and acoustic | |
DIN 12880:2007-05 , EN 61010-1 (IEC 61010-1), EN 61010-2-010 | |
Stainless Steel | |
IR Sensor | |
1 to 20% | |
18°C to 50°C | |
60 min. | |
Mains cable with plug (German type) | |
115/230 V, 50/60 Hz |
15 kg | |
TwinDISPLAY: adaptive multifunctional digital PID-microprocessor controller with 2 high-definition TFT-colour displays | |
0 to 20% | |
Textured Stainless Steel | |
40 to 97% | |
CO2 Incubator | |
2 high-definition TFT-colour displays (TwinDISPLAY) | |
Perforated Stainless Steel | |
50/60 Hz |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.