missing translation for 'onlineSavingsMsg'
Learn More

Micronova™ Cleanroom Pens
BIOPHARMA

Product Code. 11992475 Shop All Micronova Products
Change view
Click to view available options
Color:
Black
Blue
Red
Unit Size:
Pack of 10
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
11992475 Black Pack of 10
13187716 Blue Pack of 10
13197716 Red Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11992475 Supplier Micronova™ Supplier No. PEN01
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

These low-sodium, indelible ink ballpoint pens feature a unique polypropylene barrel that limits particle generation and holds up to IPA, bleach and phenol wipe downs. Available irradiated, capped or click versions, and with black, blue or red ink.

Low-sodium ink

The Micronova Cleanroom Ballpoint Pens, with low sodium ink and low particulate barrels, are designed for documentation and labeling in cleanrooms and aseptic environments. Gamma irradiated pens are suitable for Aseptic ISO 5 and other clean production areas.

  • Ball point cleanroom pens with indelible, low sodium ink
  • Designed for documentation and labeling in cleanrooms
  • White, high-density polypropylene barrel
  • Holds up to IPA, bleach and phenol wipe downs
  • Capped or Retractable options available
  • Black, blue or red ink
  • Available gamma irradiated for aseptic environments

Specifications

Material HDPE
Color Black
Description Capped
Length (English) 5.75 in.
Autoclavable Not Autoclavable
Disposable Reusable
For Use With (Application) Cleanroom and Aseptic Environment Documentation and Labeling
Product Type Cleanroom Pen
Sterility Non-sterile
Tip Style 0.7 mm Point Tip
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.