missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ChromaCare™ LC-MS Organic Rinse, Probe Wash 2, Thermo Scientific™
Shop All Thermo Scientific Products
Click to view available options
Quantity:
2.5 L
Packaging:
Amber Glass Bottle
Unit Size:
2.50 L
Description
- Suitable as a cleaning solution to clean injection needle on LC instrument
- Save time by using off-the-shelf blend -- pre-made for convenience
- Helps to maintain clean sample lines and reduce sample carryover
- Commonly used as auto-sampler cleaning solution in LC-MS
- Packaged in 2.5L amber glass bottle

Specifications
Specifications
CAS | 7732-18-5, 64-18-6, 75-05-8 |
Concentration or Composition (by Analyte or Components) | Water with 40% Acetonitrile and 0.1% Formic Acid |
Packaging | Amber Glass Bottle |
Quantity | 2.5 L |
Physical Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction