Learn More
ChromaCare™ LC-MS Instrument Flush Solution
ChromaCare™ LC-MS Instrument Flush Solution is designed to prepare LC/MS instruments for start-up. By reducing background noise, this solution facilitates instrument installation and preventative maintenance routines. | CAS: 75-05-8; 67-56-1; 67-63-0; 7732-18-5
Brand: Thermo Scientific T111101000
139.11 EUR valid until 2025-07-13
Use promo code "24135" to get your promotional price.
View more versions of this product
Description
- Designed by Thermo Scientific to maximize cleaning while preserving instrument integrity in Vanquish™ and UltiMate™ UHPLC systems with QQQ MS
- Comprised of a unique blend of polar protic and aprotic solvents for removing a wide variety of contaminants
- Particularly suitable for new LC/MS and UHPLC/MS installations to rapidly stabilize baseline
- A blend of 25% acetonitrile, 25% methanol, 25%, water, and 25% 2-propanol (IPA)
- Offered in 1L borosilicate bottles and 2.5L amber glass bottles

Specifications
LC-MS Instrument Flush Solution | |
75-05-8; 67-56-1; 67-63-0; 7732-18-5 | |
Colorless | |
25% Acetonitrile, 25% Methanol, 25% Water, 25% 2-Propanol (IPA) |
Borosilicate glass bottle | |
Liquid | |
1 L | |
LC-MS |
Safety and Handling
- ChromaCare LC/MS Instrument Flush Solution
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity after single exposure Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
- H331-Toxic if inhaled.
- H336-May cause drowsiness or dizziness.
- H370-Causes damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P311-Call a POISON CENTER/doctor/ .
Supplemental information
- MIXTURE LIST-Contains : Acetonitrile, Methanol, isopropanol
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only