missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell™ Microflex™ 93-260 Chemical-Resistant Disposable Glove

Provides thin yet tough chemical protection with unparalleled comfort. Microflex™ 93-260 Disposable Gloves have a solvent resistant exterior, an acid and base resistant middle layer and an easy donning interior layer.
28.70 - 29.70
Specificatie
Certificeringen/conformiteit | ASTM D3577, TYPE II, PPE 89/686, EN 420, 388 & 374 CATEGORY III |
---|---|
Kleur | Green, Blue |
Voor eenmalig gebruik | Single-use |
Hand | Ambidextrous |
Materiaal | Nitrile, Neoprene |
Productcode | Merk | Maat | Prijs | Hoeveelheid en beschikbaarheid | |||||
---|---|---|---|---|---|---|---|---|---|
Productcode | Merk | Maat | Prijs | Hoeveelheid en beschikbaarheid | |||||
15623074
|
Ansell™
93260060 |
X-Small |
28.70
|
![]() |
|||||
15633074
|
Ansell™
93260070 |
Small |
28.70
50 stuks |
![]() |
|||||
15643074
|
Ansell™
93260080 |
Medium |
28.70
50 stuks |
![]() |
|||||
15653074
|
Ansell™
93260090 |
Large |
28.70
|
![]() |
|||||
15663074
|
Ansell™
93260100 |
X-Large |
29.70
|
![]() |
|||||
15613074
![]() |
Ansell™
93260110 |
2X-Large |
28.70
50 stuks |
![]() |
|||||
Beschrijving
- Three layer design for superior protection against harsh chemicals including acids, solvents and bases
- Thin mil construction provides enhanced tactility and dexterity
- Extra soft material and ergonomic design for outstanding fit, feel and flexibility for longer wear time
- Lower acceptable pinhole rate (0.65 AQL) and extended cuff for reliable protection against hazardous substances
- Silicone free formulation and processing ensure better product protection
SIZE CHART
- XS – 5.5 to 6
- S – 6.5 to 7
- M – 7.5 to 8
- L – 6.5 to 9
- XL – 9.5 to 10
- 2XL – 10.5 to 11
Specificatie
ASTM D3577, TYPE II, PPE 89/686, EN 420, 388 & 374 CATEGORY III | |
Single-use | |
Nitrile, Neoprene | |
Disposable Gloves | |
Textured | |
93-260 | |
0.65 AQL | |
Harsh chemicals including acids, solvents and bases | |
300 mm |
Green, Blue | |
Ambidextrous | |
Powder Free | |
0.198 mm | |
Extended, Beaded | |
Dispenser box | |
50Pack | |
22 MPa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title