missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell™ ChemTek™ 38-520 Series Butyl Polymer Immersion Gloves
Provides the best resistance to the most aggressive chemicals without compromising dexterity or comfort. ChemTekTM gloves are surprisingly flexible and provide excellent dexterity.
Brand: Ansell™ 38520080
61.50 EUR valid until 2025-10-31
Use promo code "24133" to get your promotional price.
Description
- Smooth finish in black
- Easy donning and good grip
- Maximum protection from hazardous chemicals
- Surprisingly flexible and provide excellent dexterity
- Designed for comfort with a natural, curved ergonomic shape and soft feel
- Absence of latex proteins means there's no risk of latex allergies
- Ideal for the chemical industry, oil and petrol refining, printing industry, automotive industry, mining and aerospace
Specifications
| Butyl | |
| Industrial | |
| Black | |
| Yes | |
| Immersion Gloves | |
| 20 mil | |
| Black | |
| Black | |
| Individually wrapped bags with insert | |
| Highly chemical resistance |
| Butyl | |
| Butyl Polymer | |
| Yes | |
| Butyl Polymer | |
| 8 | |
| 0.0268 in. | |
| Beaded/Rolled | |
| Yes | |
| 1 Pair | |
| 38-520 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction