Learn More
Macherey-Nagel™ Cellulose Extraction Thimbles
Minimizes the hands-on time involved in extraction. Macherey Nagel™ Cellulose Extraction Thimbles are used for holding solid materials from which substances are to be extracted with a suitable solvent.
Brand:  Macherey-Nagel™ 645004
 View more versions of this product
Product Code. 10536101
  • ONLINE EXCLUSIVE PRICE
    € 97.20
    € 94.80 / Pack of 25
    Save € 2.40
    2% Off
Only null left
Estimated Shipment: 12-11-2025
Add to basket
This item is not returnable. View return policy
Description
Description
  • Manufactured from pure cellulose
  • Used in the fields of air and waste gas analysis for collecting solid particles
  • The choice of analytical chemists using Soxhlet or automated extractors
Specifications
Specifications
25 Pk.
Cellulose
100 mm
15 mm
Analysis of soil contaminants, Analysis of soil contaminants
Extraction Thimbles
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.