missing translation for 'onlineSavingsMsg'
Learn More

Corning™ DMEM (Dulbecco's Modified Eagle's Medium)
BIOPHARMA

Product Code. p-8007943
Change view
Click to view available options
Quantity:
6 x 500 mL
6 x 1 L
1 x 10 L
1 x 20 L
With Additives:
Glucose
Glucose, Glutamine
Glucose, Glutamine, Sodium Pyruvate
Glucose, Sodium Pyruvate
Glucose, Sodium Pyruvate, HEPES
Glutamine, Glucose
Without Additives:
No Glutamine
No Glutamine, No Phenol Red
No Glutamine, No Sodium Pyruvate
No Sodium Pyruvate
None
Product Type:
DMEM Media, 1 g/L Glucose
DMEM Media, 4.5 g/L Glucose
Unit Size:
10 L
20 L
Pack of 6
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity With Additives Without Additives Product Type unitSize
15303601 6 x 1 L Glucose, Glutamine, Sodium Pyruvate None DMEM Media, 1 g/L Glucose Pack of 6
15323601 6 x 1 L Glutamine, Glucose No Sodium Pyruvate DMEM Media, 4.5 g/L Glucose Pack of 6
15323531 6 x 500 mL Glucose, Glutamine, Sodium Pyruvate None DMEM Media, 4.5 g/L Glucose Pack of 6
15373591 6 x 1 L Glucose, Glutamine, Sodium Pyruvate None DMEM Media, 4.5 g/L Glucose Pack of 6
15353531 6 x 500 mL Glucose, Glutamine No Sodium Pyruvate DMEM Media, 4.5 g/L Glucose Pack of 6
15353551 6 x 500 mL Glucose, Sodium Pyruvate No Glutamine DMEM Media, 4.5 g/L Glucose Pack of 6
15383551 6 x 500 mL Glucose No Glutamine, No Sodium Pyruvate DMEM Media, 4.5 g/L Glucose Pack of 6
15393551 6 x 500 mL Glucose, Sodium Pyruvate, HEPES No Glutamine DMEM Media, 4.5 g/L Glucose Pack of 6
15383561 6 x 500 mL Glucose, Sodium Pyruvate No Glutamine, No Phenol Red DMEM Media, 4.5 g/L Glucose Pack of 6
15393611 6 x 1 L Glucose, Sodium Pyruvate No Glutamine DMEM Media, 4.5 g/L Glucose Pack of 6
15393591 1 x 10 L Glucose, Glutamine, Sodium Pyruvate None DMEM Media, 4.5 g/L Glucose 10 L
15313621 1 x 10 L Glucose, Sodium Pyruvate No Glutamine DMEM Media, 4.5 g/L Glucose 10 L
15303621 1 x 20 L Glucose, Sodium Pyruvate No Glutamine DMEM Media, 4.5 g/L Glucose 20 L
15333621 6 x 1 L Glucose, Sodium Pyruvate, HEPES No Glutamine DMEM Media, 4.5 g/L Glucose Pack of 6
15323621 6 x 1 L Glucose No Glutamine, No Sodium Pyruvate DMEM Media, 4.5 g/L Glucose Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
This item is not returnable. View return policy
Product Code. 15303601 Supplier Corning™ Supplier No. 10014CM

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Corning™ DMEM (Dulbecco's Modified Eagle's Medium) support cell culture growth and substitution of Pyridoxine HCl for Pyridoxal ensures improved stability over original formulation. Products undergo a variety of tests to determine functionality and toxicity using growth-promotion analysis.

  • Modification of Basal Medium Eagle (BME) containing four-fold concentrations of the amino acids and vitamins found in BME
  • Original formulation was used to culture embryonic mouse cells
  • Has been modified to support variety of normal and transformed cells, such as primary cultures of mouse and chicken cells
  • Formulations can differ in amount or absence of components such as glucose, sodium pyruvate, sodium bicarbonate and L-glutamine
  • Tested to determine pH and osmolality
  • Low levels of endotoxin, and absence of contamination
  • Tested for bacterial endotoxin using the Limulus Amoebocyte Lysate (LAL) Chromogenic Assay
  • Osmolality: 335±mOsm/kg H2O
  • Mycoplasma: Tested Negative
  • Sterility: Pass
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 X
Form Liquid
Product Type DMEM Media, 1 g/L Glucose
With Additives Glucose, Glutamine, Sodium Pyruvate
Without Additives None
For Use With (Application) To assess growth promotion and cytotoxicity characteristics
Quantity 6 x 1 L
Endotoxin Level <0.25EU/mL
Formulation Inorganic Salts: 200mg/L CaCl2 (anhydrous), 0.1mg/L Fe(NO3)3.9H2O, 400mg/L KCl, 97.7mg/L MgSO4 (anhydrous), 6400mg/L NaCl, 125mg/L NaH2PO4.H2O, 3700mg/L NaHCO3; Amino Acids: 84mg/L L-Arginine. HCl, 62.57mg/L L-Cystine.2HCl, 584mg/L L-glutamine, 30mg/L Glycine, 42mg/L L-Histidine.HCl.H2O, 104.8mg/L L-Isoleucine, 104.8mg/L L-Leucine, 146.2mg/L L-Lysine. HCl, 30mg/L L-Methionine, 66mg/L L-Phenylalanine, 42mg/L L-Serine, 95.2mg/L L-Threonine, 16mg/L L-Tryptophan, 103.79mg/L L-Tyrosine.2Na.2H2O, 20mg/L L-Valine; Vitamins: 4mg/L D-Calcium pantothenate, 4mg/L Choline chloride, 4mg/L Folic Acid, 7.2mg/L i-Inositol, 4mg/L Nicotinamide, 4mg/L Pyridoxine. HCl, 0.4mg/L Riboflavin, 4mg/L Thiamine. HCl; Other: 1000mg/L D-Glucose, 15mg/L Phenol red. Na, 110mg/L Sodium pyruvate.
Content And Storage 2 to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.