Learn More
Cole-Parmer™ Essentials Replacement Cell Culturing Flask
Description
- Constructed of borosilicate glass
The culture vessels incorporate a unique base design which, together with the bulb-ended stirrer, ensures the cells are lifted into suspension at the lowest possible speeds. This gentle stirring action promotes high cell yields by preventing cell shearing. Replacement flasks do not include caps or stirring mechanisms.
Specifications
Specifications
| Capacity (Metric) | 1000 mL |
| Material | Borosilicate Glass |
| For Use With (Equipment) | Biological Stirrers Glass no End Caps, Seals or Stirrer Rods |
| Bottom Shape | Flat |