Learn More
Fisher Chemical Buffer Solution, Colour Coded Red, pH 4, Traceable to NIST, 250ml
Brand:  Fisher Chemical J/2826/07
25.03 EUR valid until 2026-02-28
BEST PRICE on Promo! Use promo code "28012" to get your promotional price.
Buffer Solution, Colour Coded Red, pH 4, Traceable to NIST, 250ml
Product Code. 15840064
  • 26.35€ / 0.25 L
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.