Learn More
BUCHI Calibration and Verification Standards for BUCHI Melting Point Instruments
Utilize the embedded firmware and software on your Melting Point apparatus to calibrate and verify your instrument at any time.
Brand: BUCHI 11055019
View more versions of this product
Description
Utilize the embedded firmware and software on your BUCHI Melting Point apparatus to calibrate and verify your instrument at any time.
- Comprehensive IQ/OQ procedures including certified calibration and verification compounds for GLP/GMP laboratories
- Guided calibrations and verifications provide easy and reliable results
- Ensures compliance from 10°C to 400°C
Specifications
| Verification Kit | |
| BUCHI Melting Point Instrument | |
| Three certified standards |
Safety and Handling
- Verification Set M-560/565
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Carcinogenicity Category 1A
Hazard Statement
- H302-Harmful if swallowed.
- H350-May cause cancer.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P270-Do not eat, drink or smoke when using this product.
- P330-Rinse mouth.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.,Store locked up.
- P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..,Dispose of contents/container to in accordance with local/regional/national/international regulations..
Your input is important to us. Please complete this form to provide feedback related to the content on this product.