missing translation for 'onlineSavingsMsg'
Learn More

BUCHI Calibration and Verification Standards for BUCHI Melting Point Instruments

Product Code. 12053303 Shop All Buchi Products
Click to view available options
Product Type:
Calibration Set
Verification Kit
For Use With (Equipment):
BUCHI Melting Point Instrument
Unit Size:
Each
This item is not returnable. View return policy
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
This item is not returnable. View return policy
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.

Utilize the embedded firmware and software on your Melting Point apparatus to calibrate and verify your instrument at any time.

Utilize the embedded firmware and software on your BUCHI Melting Point apparatus to calibrate and verify your instrument at any time.

  • Comprehensive IQ/OQ procedures including certified calibration and verification compounds for GLP/GMP laboratories
  • Guided calibrations and verifications provide easy and reliable results
  • Ensures compliance from 10°C to 400°C

Specifications

Product Type Calibration Set
For Use With (Equipment) BUCHI Melting Point Instrument
compliance-icons
Product Identifier
  • Calibration Set M-560/565
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Acute toxicity Category 4
  • Germ cell mutagenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Carcinogenicity Category 1B
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.,Causes skin irritation.
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.,Causes serious eye irritation.,Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.,Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.,Wear protective gloves/protective clothing/eye protection/face protection.,Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.,IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.,IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.,If skin irritation occurs: Get medical advice/attention.,If skin irritation occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.,Take off contaminated clothing and wash it before reuse.,Take off contaminated clothing and wash it before reuse.,Take off contaminated clothing and wash it before reuse.
  • P337+P313-If eye irritation persists: Get medical advice/attention.,If eye irritation persists: Get medical advice/attention.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.