missing translation for 'onlineSavingsMsg'
Learn More

BUCHI Calibration and Verification Standards for BUCHI Melting Point Instruments

Utilize the embedded firmware and software on your Melting Point apparatus to calibrate and verify your instrument at any time.

Brand:  BUCHI 037454


 View more versions of this product

Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.

Product Code. 10532511

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Includes: Vanillin, Caffeine, Phenacetin
This item is not returnable. View return policy

Description

Description

Utilize the embedded firmware and software on your BUCHI Melting Point apparatus to calibrate and verify your instrument at any time.

  • Comprehensive IQ/OQ procedures including certified calibration and verification compounds for GLP/GMP laboratories
  • Guided calibrations and verifications provide easy and reliable results
  • Ensures compliance from 10°C to 400°C
Specifications

Specifications

Calibration Kit
BUCHI B-540/545 Melting Point apparatus
Vanillin, caffeine, phenacetin
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Calibration Kit for Melting Point

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Acute toxicity Category 4
  • Oxidising solid Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Serious eye damage/eye irritation Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2

Hazard Statement
  • H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
  • H315-Causes skin irritation.
  • H302-Harmful if swallowed.
  • H272-May intensify fire; oxidiser.
  • H373-May cause damage to organs.
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P264-Wash thoroughly after handling.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P321-Specific treatment (see on this label).
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P270-Do not eat, drink or smoke when using this product.
  • P220-Keep/Store away from clothing/combustible materials.
  • P330-Rinse mouth.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P361 + P364-Take off immediately all contaminated clothing and wash it before reuse.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
  • P405-Store locked up.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P370+P378-In case of fire: Use CO2, extinguishing powder or water spray. to extinguish.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.