Learn More
Proteintech Broad Range Prestained Protein Marker
The PL00002 Broad Range Protein Marker is a ready-to-use three-color protein standard with 13 prestained proteins covering a wide range of molecular weights from 3 to 245 kDa in Tris-Glycine buffer.
Brand:  Proteintech PL00002-500UL
Product Code. 17318262
  • 962.46 DKK / 500µL
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
  • The PL00002 Broad Range Prestained Protein Marker is designed for monitoring protein separation during SDS-polyacrylamide gel electrophoresis, verification of Western transfer efficiency on membranes (nitrocellulose, PVDF, or nylon) and for estimating the size of proteins.
  • Contents: Approximately 0.1~0.4 mg/mL of each protein in the buffer (20 mM Tris-phosphate (pH 7.5 at 25°C), 2 % SDS, 0.2 mM Dithiothreitol, 3.6 M Urea, and 15 % (v/v) Glycerol).
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.